Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000029.3 | NP_000020.1 | 4, 5 | 1785 | ||
GenBank mRNA | AB209769.1 | - | 4, 5 | 1297 | ||
AK222798.1 | - | 4, 5 | 1316 | |||
AK222815.1 | - | 4, 5 | 1317 | |||
AK293507.1 | - | 5, 6 | 1070 | |||
AK303755.1 | - | 5, 6 | 1258 | |||
AK307978.1 | - | 5, 6 | 1231 | |||
AK312291.1 | - | 4, 5 | 1395 | |||
BC011519.1 | - | 4, 5 | 1305 | 4213559 | ||
BT006851.1 | - | 3, 4 | 1277 | |||
K02215.1 | - | 4, 5 | 1316 |