Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144860.2 | NP_659109.2 | 13, 14 | 2650 | ||
GenBank mRNA | AK053035.1 | - | 6, 7 | 873 | ||
AK080847.1 | - | 7, 8 | 926 | |||
AY147848.1 | - | 13, 14 | 2149 | |||
AY149907.1 | - | 13, 14 | 1970 | |||
AY245539.1 | - | 13, 14 | 2650 | |||
AY974091.1 | - | 13, 14 | 1943 | |||
BC011287.1 | - | 2, 3 | 247 | |||
BC083072.1 | - | 3, 4 | 328 | |||
BC131662.1 | - | 14, 15 | 2116 | 40110696 |