Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145413.4 | NP_663388.1 | 2 | 292 | ||
GenBank mRNA | AK034081.1 | - | 3 | 399 | ||
AK049705.1 | - | 1 | 94 | |||
AK132149.1 | - | 2 | 293 | |||
BC019381.1 | - | 2 | 311 | 4206622 | ||
BC023737.1 | - | 3 | 508 | 5352660 | ||
BC024412.1 | - | 1 | 242 | 4971483 | ||
BC031473.1 | - | 2 | 404 | 3990487 |