Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olfr424 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146721.2 | NP_666932.2 | 1 | 277 | ||
GenBank mRNA | BC104096.1 | - | 1 | 277 | 40043031 | |
BC104097.1 | - | 1 | 277 | 40043033 |