Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pilra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153510.3 | NP_705730.1 | 3, 4 | 835 | ||
GenBank mRNA | AJ400844.1 | - | 733 | |||
AK041413.1 | - | 2, 3 | 1126 | |||
AK041670.1 | - | 3, 4 | 747 | |||
AK170000.1 | - | 3, 4 | 747 | |||
BC115565.2 | - | 720 | 40102015 |