Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adamts4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172845.2 | NP_766433.1 | 1 | 597 | ||
GenBank mRNA | AK028364.1 | - | 597 | |||
AK083534.1 | - | 1 | 597 | |||
AK157287.1 | - | 598 | ||||
AK164900.1 | - | 1 | 593 | |||
AK173001.1 | - | 477 | ||||
BC027773.1 | - | 1 | 231 | 5345809 |