Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam221b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175517.3 | NP_780726.1 | 3 | 1216 | ||
GenBank mRNA | AK038644.1 | - | 3 | 1503 | ||
AK076646.1 | - | 3 | 1216 | |||
AK076680.1 | - | 3 | 1164 | |||
BC119804.1 | - | 3 | 1209 | 40107542 | ||
BC122881.1 | - | 3 | 1209 | 40107552 |