Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003683.5 | NP_003674.1 | 2, 3 | 316 | ||
XM_017028485.1 | XP_016883974.1 | 2, 3 | 307 | |||
XM_017028486.1 | XP_016883975.1 | 2, 3 | 302 | |||
GenBank mRNA | AK223185.1 | - | 2, 3 | 277 | ||
AK223215.1 | - | 2, 3 | 273 | |||
AK303000.1 | - | 2, 3 | 276 | |||
AK313116.1 | - | 2, 3 | 273 | |||
BC000380.2 | - | 2, 3 | 263 | 2820455 | ||
BC014787.1 | - | 2, 3 | 238 | |||
BG829556.1 | - | 304 | ||||
DA275615.1 | - | 2, 3 | 318 | |||
U79775.1 | - | 2, 3 | 261 |