Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP1R16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172735.2 | NP_001166206.1 | 5 | 752 | ||
NM_015568.3 | NP_056383.1 | 5 | 752 | |||
XM_011528768.2 | XP_011527070.1 | 4 | 1315 | |||
GenBank mRNA | AB020630.2 | - | 5 | 740 | ||
AB177855.1 | - | 4 | 379 | |||
AF362910.1 | - | 4 | 603 | |||
BC131801.1 | - | 5 | 709 | 40081818 | ||
BC152467.1 | - | 5 | 740 | 8862603 |