Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: INTU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015693.3 | NP_056508.2 | 8 | 1481 | ||
XM_011531844.2 | XP_011530146.1 | 8 | 1505 | |||
XM_011531845.2 | XP_011530147.1 | 8 | 1508 | |||
XM_011531846.1 | XP_011530148.1 | 8 | 1453 | |||
XM_011531847.1 | XP_011530149.1 | 8 | 890 | |||
XM_011531848.2 | XP_011530150.1 | 3 | 316 | |||
XM_011531849.2 | XP_011530151.1 | 8 | 1508 | |||
XM_011531850.2 | XP_011530152.1 | 8 | 1508 | |||
XM_011531851.2 | XP_011530153.1 | 8 | 1508 | |||
XM_017008025.1 | XP_016863514.1 | 5 | 424 | |||
XM_017008026.1 | XP_016863515.1 | 8 | 1508 | |||
GenBank mRNA | AB033110.1 | - | 8 | 1448 | ||
AK123320.1 | - | 1 | 821 | |||
AK291481.1 | - | 8 | 1463 | |||
BC051698.1 | - | 10 | 3949 | |||
BC130611.1 | - | 8 | 1447 | 40146642 | ||
HQ234311.1 | - | 389 |