Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POLM SIRNA Assays ›
Gene Symbol:
POLM
Gene Name:
DNA polymerase mu
Gene Aliases:
Pol Mu, Tdt-N
Chromosome Location:
Chr.7: 44072062 - 44082540 on Build GRCh38
UniGene ID:
Hs.596982
Species:
Human
siRNA ID
HSS120686
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POLM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284330.1
NP_001271259.1
2
404
NM_001284331.1
NP_001271260.1
2
404
NM_013284.3
NP_037416.1
2
404
NR_104299.1
2
404
XM_005249711.2
XP_005249768.1
2
375
XM_006715692.2
XP_006715755.1
2
375
XM_006715696.2
XP_006715759.1
2
375
XM_011515275.1
XP_011513577.1
2
375
XM_011515278.1
XP_011513580.1
2
375
XM_011515279.1
XP_011513581.1
2
375
XM_011515286.2
XP_011513588.1
2
375
XM_011515287.2
XP_011513589.1
2
375
XM_017011998.1
XP_016867487.1
2
375
XM_017011999.1
XP_016867488.1
2
375
XM_017012000.1
XP_016867489.1
2
375
XM_017012001.1
XP_016867490.1
2
375
XM_017012002.1
XP_016867491.1
2
357
XM_017012003.1
XP_016867492.1
2
375
XM_017012004.1
XP_016867493.1
2
375
XM_017012005.1
XP_016867494.1
2
375
GenBank mRNA
AF176097.1
-
2
344
AJ131891.2
-
2
347
AK023002.1
-
2
379
AK092903.1
-
2
354
AK291638.1
-
2
348
AK294449.1
-
2
375
AK307634.1
-
2
347
AU309799.1
-
2
404
BC026306.1
-
2
375
4813399
BC049202.1
-
2
349
5246144
BC062590.1
-
2
328
5215523
DC407437.1
-
2
394
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-directed DNA polymerase
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via nonhomologous end joining
DNA recombination
somatic hypermutation of immunoglobulin genes
B cell differentiation
DNA biosynthetic process
DNA binding
DNA-directed DNA polymerase activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-9xb8z:80/10.239.165.207:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0