Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FKBP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322963.1 | NP_001309892.1 | 4 | 381 | ||
NM_001322964.1 | NP_001309893.1 | 3 | 200 | |||
NM_004116.4 | NP_004107.1 | 3 | 313 | |||
NM_054033.3 | NP_473374.1 | 3 | 313 | |||
NR_136536.1 | 5 | 470 | ||||
NR_136538.1 | 4 | 402 | ||||
NR_136539.1 | 3 | 200 | ||||
XM_011532699.2 | XP_011531001.1 | 3 | 262 | |||
XM_017003593.1 | XP_016859082.1 | 5 | 416 | |||
XM_017003594.1 | XP_016859083.1 | 3 | 180 | |||
XM_017003595.1 | XP_016859084.1 | 2 | 161 | |||
XM_017003596.1 | XP_016859085.1 | 4 | 407 | |||
GenBank mRNA | AF322070.1 | - | 3 | 205 | ||
BC002614.1 | - | 3 | 256 | 3161896 | ||
BF216711.1 | - | 3 | 167 | |||
BF969580.1 | - | 3 | 300 | |||
BI546013.1 | - | 3 | 278 | |||
BM802489.1 | - | 3 | 313 | |||
BU147063.1 | - | 3 | 151 | |||
BU508033.1 | - | 3 | 277 | |||
D38037.1 | - | 3 | 236 | |||
L37086.1 | - | 3 | 230 | |||
S69800.1 | - | 3 | 236 | |||
S69815.1 | - | 3 | 236 |