Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001448.1 | 11 | 1187 | |||
GenBank mRNA | AJ132821.1 | - | 10 | 1121 | ||
AJ132822.1 | - | 10 | 1003 | |||
AJ132823.1 | - | 9 | 937 | |||
AJ132826.1 | - | 1 | 117 | |||
AJ132827.1 | - | 1 | 117 | |||
BC047448.1 | - | 11 | 1321 | |||
BC067864.1 | - | 9 | 980 |