Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RBCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323956.1 | NP_001310885.1 | 4, 5 | 1158 | ||
NM_001323958.1 | NP_001310887.1 | 3, 4 | 1013 | |||
NM_006462.5 | NP_006453.1 | 3, 4 | 1013 | |||
NM_031229.3 | NP_112506.2 | 4, 5 | 1158 | |||
NR_136659.1 | 4, 5 | 1158 | ||||
XM_005260645.1 | XP_005260702.1 | 4, 5 | 954 | |||
XM_011529137.1 | XP_011527439.1 | 5, 6 | 1077 | |||
XM_011529138.1 | XP_011527440.1 | 5, 6 | 1032 | |||
XM_011529139.2 | XP_011527441.1 | 5, 6 | 1077 | |||
XM_011529140.1 | XP_011527442.1 | 5, 6 | 1077 | |||
XM_017027594.1 | XP_016883083.1 | 4, 5 | 1203 | |||
XM_017027595.1 | XP_016883084.1 | 4, 5 | 548 | |||
GenBank mRNA | AB107766.1 | - | 3, 4 | 344 | ||
AK297524.1 | - | 4, 5 | 904 | |||
AK303357.1 | - | 4, 5 | 873 | |||
BC000983.2 | - | 3, 4 | 708 | |||
BC015219.2 | - | 4, 5 | 871 | 3877587 | ||
U67322.1 | - | 3, 4 | 899 |