Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NXPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007225.2 | NP_009156.2 | 2 | 383 | ||
GenBank mRNA | AK308928.1 | - | 2 | 197 | ||
AL834282.1 | - | 2 | 461 | |||
AY358526.1 | - | 2 | 153 | |||
BC022541.1 | - | 2 | 202 | 4794968 |