Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031623.2 | NP_001026794.1 | 10 | 1961 | ||
NM_015555.2 | NP_056370.2 | 10 | 1961 | |||
XM_005248994.1 | XP_005249051.1 | 10 | 1981 | |||
XM_011514460.1 | XP_011512762.1 | 9 | 1720 | |||
XM_011514461.1 | XP_011512763.1 | 8 | 2336 | |||
XM_011514462.2 | XP_011512764.1 | 10 | 1981 | |||
GenBank mRNA | AB011148.1 | - | 10 | 1780 | ||
AK000983.1 | - | 1 | 467 | |||
AK225806.1 | - | 10 | 1788 | |||
AY055204.1 | - | 10 | 1962 | |||
BC021712.2 | - | 10 | 1878 | 4821000 | ||
BC098382.1 | - | 10 | 1804 | 5295845 | ||
BX648081.1 | - | 10 | 1807 |