Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Txndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289598.1 | NP_001276527.1 | 6, 7 | 826 | ||
NM_001289599.1 | NP_001276528.1 | 6, 7 | 793 | |||
NM_145367.4 | NP_663342.3 | 6, 7 | 850 | |||
GenBank mRNA | AK136046.1 | - | 6, 7 | 738 | ||
AK142049.1 | - | 6, 7 | 836 | |||
AK150781.1 | - | 6, 7 | 834 | |||
AK169680.1 | - | 6, 7 | 817 | |||
AK185996.1 | - | 2, 3 | 166 | |||
AK205431.1 | - | 2, 3 | 166 | |||
AK224911.1 | - | 5, 6 | 503 | |||
AK224913.1 | - | 6, 7 | 837 | |||
AK224922.1 | - | 6, 7 | 795 | |||
AK224923.1 | - | 6, 7 | 817 | |||
AK224924.1 | - | 6, 7 | 825 | |||
AK224926.1 | - | 6, 7 | 795 | |||
AK224927.1 | - | 6, 7 | 846 | |||
AK224933.1 | - | 6, 7 | 787 | |||
AK224934.1 | - | 6, 7 | 841 | |||
AK224936.1 | - | 6, 7 | 840 | |||
AK224937.1 | - | 6, 7 | 839 | |||
AK224938.1 | - | 6, 7 | 834 | |||
AK224939.1 | - | 6, 7 | 827 | |||
AK224943.1 | - | 6, 7 | 798 | |||
AK224945.1 | - | 6, 7 | 834 | |||
AK224946.1 | - | 6, 7 | 837 | |||
AK224947.1 | - | 6, 7 | 824 | |||
AK224948.1 | - | 6, 7 | 798 | |||
AK224949.1 | - | 6, 7 | 787 | |||
AK224950.1 | - | 6, 7 | 787 | |||
AK224951.1 | - | 6, 7 | 845 | |||
AK224955.1 | - | 1, 2 | 98 | |||
AK224956.1 | - | 6, 7 | 757 | |||
AK224957.1 | - | 6, 7 | 803 | |||
AK224958.1 | - | 6, 7 | 803 | |||
AK224960.1 | - | 6, 7 | 845 | |||
AK224962.1 | - | 6, 7 | 805 | |||
AK224963.1 | - | 6, 7 | 846 | |||
AK224964.1 | - | 6, 7 | 846 | |||
AK224965.1 | - | 6, 7 | 822 | |||
AK224966.1 | - | 6, 7 | 822 | |||
AK224967.1 | - | 6, 7 | 813 | |||
AK224968.1 | - | 4, 5 | 380 | |||
AK224969.1 | - | 4, 5 | 380 | |||
AK224970.1 | - | 6, 7 | 787 | |||
AK224971.1 | - | 6, 7 | 847 | |||
AK224972.1 | - | 6, 7 | 837 | |||
AK224973.1 | - | 6, 7 | 787 | |||
AK224974.1 | - | 6, 7 | 790 | |||
AK224975.1 | - | 1, 2 | 25 | |||
AK224976.1 | - | 6, 7 | 820 | |||
AK224977.1 | - | 6, 7 | 834 | |||
AK224978.1 | - | 6, 7 | 834 | |||
AK224979.1 | - | 6, 7 | 821 | |||
AK224981.1 | - | 6, 7 | 791 | |||
AK224982.1 | - | 6, 7 | 846 | |||
AK224983.1 | - | 6, 7 | 835 | |||
AK224984.1 | - | 6, 7 | 800 | |||
AK224985.1 | - | 6, 7 | 804 | |||
AK224986.1 | - | 6, 7 | 848 | |||
AK224988.1 | - | 6, 7 | 824 | |||
AK224989.1 | - | 6, 7 | 841 | |||
AK224990.1 | - | 6, 7 | 836 | |||
AK224991.1 | - | 6, 7 | 831 | |||
AK224992.1 | - | 6, 7 | 831 | |||
AK224993.1 | - | 6, 7 | 831 | |||
AK224997.1 | - | 6, 7 | 798 | |||
AK224998.1 | - | 6, 7 | 796 | |||
AK224999.1 | - | 6, 7 | 818 | |||
AK225004.1 | - | 6, 7 | 839 | |||
AK225005.1 | - | 6, 7 | 817 | |||
AK225007.1 | - | 6, 7 | 839 | |||
AK225010.1 | - | 6, 7 | 839 | |||
AK225011.1 | - | 6, 7 | 839 | |||
AK225012.1 | - | 6, 7 | 608 | |||
AK225013.1 | - | 6, 7 | 608 | |||
AK225014.1 | - | 6, 7 | 817 | |||
AK225015.1 | - | 6, 7 | 608 | |||
AK225016.1 | - | 6, 7 | 747 | |||
AK225017.1 | - | 6, 7 | 795 | |||
AK225019.1 | - | 6, 7 | 827 | |||
AK225020.1 | - | 6, 7 | 778 | |||
AK225021.1 | - | 6, 7 | 778 | |||
AK225022.1 | - | 6, 7 | 626 | |||
AK225023.1 | - | 6, 7 | 852 | |||
AY243534.1 | - | 6, 7 | 813 | |||
AY548112.1 | - | 6, 7 | 774 | |||
BC016252.1 | - | 6, 7 | 817 | 4911455 | ||
BC024505.1 | - | 6, 7 | 762 | 4983496 | ||
BC036155.1 | - | 5, 6 | 436 | |||
BC046789.1 | - | 6, 7 | 776 | 5716897 |