Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164071.1 | NP_001157543.1 | 6 | 709 | ||
NM_001164072.1 | NP_001157544.1 | 5 | 501 | |||
NM_011529.2 | NP_035659.1 | 5 | 500 | |||
GenBank mRNA | AK031559.1 | - | 5 | 463 | ||
AK088817.1 | - | 5 | 447 | |||
AK147727.1 | - | 6 | 655 | |||
AK165420.1 | - | 5 | 457 | |||
AK165994.1 | - | 5 | 432 | |||
BB660164.1 | - | 5 | 502 | |||
BC026148.1 | - | 5 | 482 | 3492937 | ||
BC033282.1 | - | 6 | 524 | |||
BC052685.1 | - | 5 | 401 | |||
U51907.1 | - | 5 | 439 | |||
U59864.1 | - | 5 | 513 |