Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Papolg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172555.2 | NP_766143.2 | 19 | 2209 | ||
GenBank mRNA | AK035810.1 | - | 19 | 2192 | ||
BC019635.1 | - | 4 | 562 | |||
BC023030.1 | - | 4 | 546 | |||
BC037439.1 | - | 4 | 594 | |||
BC049163.1 | - | 19 | 2153 | |||
BC059271.1 | - | 19 | 2203 | 2811338 | ||
DQ103505.1 | - | 19 | 2015 |