Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017401.2 | NP_059097.2 | 8 | 1246 | ||
GenBank mRNA | AF176098.1 | - | 8 | 1019 | ||
AJ251804.2 | - | 8 | 990 | |||
AK179467.1 | - | 1 | 25 | |||
BC056213.1 | - | 8 | 1038 | 30024844 | ||
BC056996.1 | - | 5 | 2893 |