Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301176.1 | NP_001288105.1 | 12 | 1119 | ||
NM_025518.4 | NP_079794.1 | 13 | 1163 | |||
GenBank mRNA | AK009391.1 | - | 13 | 1135 | ||
AK077451.1 | - | 12 | 1091 | |||
BC058431.1 | - | 4 | 434 | |||
BC116934.1 | - | 13 | 1081 | 40126253 | ||
BC116936.1 | - | 13 | 1081 | 40126255 |