Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mrgprb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205810.4 | NP_991379.3 | 2 | 549 | ||
GenBank mRNA | AK148044.1 | - | 2 | 545 | ||
AK148046.1 | - | 2 | 453 | |||
AK148052.1 | - | 2 | 453 | |||
AK148074.1 | - | 2 | 550 | |||
AK148108.1 | - | 2 | 454 | |||
AK148114.1 | - | 2 | 550 | |||
AK148166.1 | - | 2 | 454 | |||
AK148226.1 | - | 1 | 130 | |||
AK148241.1 | - | 2 | 454 | |||
AK148253.1 | - | 2 | 545 | |||
AK148256.1 | - | 2 | 545 | |||
AK148273.1 | - | 2 | 522 | |||
AK148279.1 | - | 2 | 503 | |||
AK148388.1 | - | 2 | 454 | |||
BC116395.1 | - | 1 | 375 | |||
BC116396.1 | - | 1 | 370 |
Set Membership: |
![]() |