Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adamts4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172845.2 | NP_766433.1 | 7 | 2272 | ||
GenBank mRNA | AK028364.1 | - | 2273 | |||
AK076295.1 | - | 6 | 1647 | |||
AK083534.1 | - | 7 | 2272 | |||
AK156429.1 | - | 5 | 855 | |||
AK157287.1 | - | 2273 | ||||
AK164900.1 | - | 7 | 2268 |