Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Limd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172397.3 | NP_765985.1 | 4, 5 | 379 | ||
GenBank mRNA | AK012581.1 | - | 4, 5 | 353 | ||
AK032430.1 | - | 4, 5 | 380 | |||
AK040486.1 | - | 4, 5 | 380 | |||
AK049809.1 | - | 4, 5 | 349 | |||
AK167349.1 | - | 4, 5 | 349 | |||
AK169829.1 | - | 4, 5 | 350 | |||
AK171794.1 | - | 4, 5 | 658 | |||
AK188791.1 | - | 1, 2 | 78 | |||
BC068130.1 | - | 4, 5 | 323 | 6413206 |