Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pdx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008814.3 | NP_032840.1 | 1 | 467 | ||
GenBank mRNA | AK020261.1 | - | 472 | |||
BC103572.1 | - | 1 | 467 | 40045668 | ||
BC103581.1 | - | 1 | 467 | 40045664 | ||
BC103582.1 | - | 1 | 467 | 40045666 | ||
BC105642.1 | - | 1 | 467 | 40045665 | ||
X74342.1 | - | 1 | 638 |