Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sfmbt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198808.1 | NP_001185737.1 | 20 | 2570 | ||
NM_177386.5 | NP_796360.2 | 20 | 2594 | |||
GenBank mRNA | AK147557.1 | - | 20 | 2575 | ||
AK147651.1 | - | 20 | 2449 | |||
AK166810.1 | - | 8 | 972 | |||
AK220408.1 | - | 17 | 2324 | |||
AY217748.1 | - | 8 | 1046 | |||
BC139354.1 | - | 19 | 2382 | 8862376 | ||
BC145610.1 | - | 19 | 2437 | 9054176 |