Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ndufa13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023312.2 | NP_075801.1 | 5 | 1091 | ||
GenBank mRNA | AF286698.1 | - | 4 | 326 | ||
AK002999.1 | - | 4 | 330 | |||
AK003012.1 | - | 4 | 344 | |||
AK010517.1 | - | 4 | 330 | |||
AK012422.1 | - | 4 | 330 | |||
BC034899.1 | - | 5 | 1091 | 1447015 | ||
BC037149.1 | - | 4 | 334 | 5149812 | ||
BC055435.1 | - | 4 | 340 | 4219489 |