Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cdkn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031550.1 | NP_113738.1 | 2 | 406 | ||
GenBank mRNA | AF474974.1 | - | 447 | |||
AF474975.1 | - | 447 | ||||
AF474976.1 | - | 2 | 382 | |||
AF474977.1 | - | 2 | 373 | |||
AY679727.1 | - | 447 | ||||
AY679728.1 | - | 2 | 382 | |||
L81167.1 | - | 2 | 406 |