Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Il1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013123.3 | NP_037255.3 | 8 | 1012 | ||
XM_006244746.2 | XP_006244808.1 | 9 | 3121 | |||
XM_006244747.2 | XP_006244809.1 | 9 | 1378 | |||
XM_006244748.2 | XP_006244810.1 | 8 | 1282 | |||
XM_006244749.2 | XP_006244811.1 | 10 | 1201 | |||
XM_006244750.2 | XP_006244812.1 | 10 | 3193 | |||
XM_006244751.2 | XP_006244813.1 | 9 | 1112 | |||
XM_006244752.2 | XP_006244814.1 | 9 | 1354 | |||
XM_006244753.2 | XP_006244815.1 | 9 | 2473 | |||
XM_006244754.2 | XP_006244816.1 | 8 | 3025 | |||
XM_008766993.1 | XP_008765215.1 | 9 | 1129 | |||
GenBank mRNA | AB079118.1 | - | 6 | 706 | ||
AB112436.1 | - | 10 | 1146 | |||
AB112437.1 | - | 8 | 1012 | |||
AB112438.1 | - | 9 | 1084 | |||
M95578.1 | - | 9 | 1333 |