Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc46a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024968.1 | NP_001020139.1 | 2 | 381 | ||
XM_006248833.2 | XP_006248895.1 | 2 | 209 | |||
GenBank mRNA | BC091179.1 | - | 2 | 381 | 7325773 |