Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ssbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053358.1 | NP_445810.1 | 14 | 1255 | ||
XM_006238532.2 | XP_006238594.1 | 15 | 1437 | |||
XM_006238533.2 | XP_006238595.1 | 15 | 1412 | |||
XM_006238534.2 | XP_006238596.1 | 15 | 1394 | |||
XM_006238535.2 | XP_006238597.1 | 15 | 1368 | |||
XM_006238536.2 | XP_006238598.1 | 14 | 1354 | |||
XM_006238537.2 | XP_006238599.1 | 14 | 1328 | |||
XM_006238538.2 | XP_006238600.1 | 14 | 1310 | |||
XM_006238539.2 | XP_006238601.1 | 12 | 1294 | |||
XM_008763902.1 | XP_008762124.1 | 15 | 3090 | |||
XM_008763903.1 | XP_008762125.1 | 16 | 2644 | |||
XM_008763904.1 | XP_008762126.1 | 15 | 3063 | |||
XM_008763905.1 | XP_008762127.1 | 15 | 3045 | |||
XM_008763906.1 | XP_008762128.1 | 15 | 3018 | |||
XM_008763907.1 | XP_008762129.1 | 14 | 3005 | |||
XM_008763908.1 | XP_008762130.1 | 14 | 2979 | |||
XM_008763909.1 | XP_008762131.1 | 14 | 2934 | |||
GenBank mRNA | AF121893.1 | - | 14 | 1255 | ||
AY724490.1 | - | 15 | 1036 | |||
BC107666.1 | - | 14 | 1239 | 7936459 | ||
FQ230168.1 | - | 9 | 533 |