Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pygo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106447.1 | NP_001099917.1 | 3 | 720 | ||
XM_006232615.1 | XP_006232677.1 | 3 | 755 | |||
XM_006232616.2 | XP_006232678.1 | 3 | 893 | |||
GenBank mRNA | BC169054.1 | - | 3 | 846 | 7930933 | |
FQ228430.1 | - | 3 | 818 |