Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PP2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252657.1 | NP_001239586.1 | 2 | 772 | ||
NR_027694.1 | 2 | 772 | ||||
GenBank mRNA | AK058178.1 | - | 2 | 756 | ||
BC105097.1 | - | 2 | 626 | |||
BC105099.1 | - | 2 | 626 |