Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308028.1 | NP_001294957.1 | 18 | 8889 | ||
NM_001308031.1 | NP_001294960.1 | 11 | 7839 | |||
NM_005246.3 | NP_005237.2 | 20 | 9275 | |||
XM_011543276.2 | XP_011541578.1 | 12 | 8091 | |||
GenBank mRNA | AK021888.1 | - | 1 | 708 | ||
AK092320.1 | - | 1 | 1643 | |||
AK123847.1 | - | 1 | 2504 |