Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SFMBT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018039.1 | NP_001018049.1 | 21 | 7614 | ||
NM_001029880.2 | NP_001025051.1 | 21 | 7716 | |||
XM_005252551.2 | XP_005252608.1 | 21 | 7497 | |||
XM_006717490.2 | XP_006717553.1 | 19 | 7489 | |||
XM_011519611.1 | XP_011517913.1 | 21 | 7715 | |||
XM_017016466.1 | XP_016871955.1 | 21 | 7617 | |||
GenBank mRNA | AK057130.1 | - | 1 | 2834 |