- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: 1500017E21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005275.1 | - | 578 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705458.1 | - | 5 | 578 | Chr. 19 - 36614365 - 36705641 [-] on Build GRCm38 |