Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144867.2 | NP_659116.1 | 5 | 527 | ||
GenBank mRNA | AK038905.1 | - | 4 | 584 | ||
AK143220.1 | - | 5 | 511 | |||
BC019561.1 | - | 5 | 476 | 4459623 | ||
BY741062.1 | - | 5 | 528 |