Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam221a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172216.1 | NP_001165687.1 | 2 | 310 | ||
NM_172727.3 | NP_766315.1 | 2 | 310 | |||
GenBank mRNA | AK052323.1 | - | 2 | 224 | ||
AK140239.1 | - | 2 | 216 | |||
BC055818.1 | - | 2 | 238 | 4487036 | ||
BY145860.1 | - | 2 | 310 |