Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006239.2 | NP_006230.2 | 17 | 2429 | ||
XM_011532039.2 | XP_011530341.1 | 16 | 2662 | |||
GenBank mRNA | AF023456.1 | - | 17 | 2439 | ||
BX648754.1 | - | 18 | 3172 |