Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RAB3IL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271686.1 | NP_001258615.1 | 4 | 591 | ||
NM_013401.3 | NP_037533.2 | 4 | 615 | |||
XM_005274140.4 | XP_005274197.1 | 4 | 580 | |||
XM_006718647.3 | XP_006718710.1 | 4 | 582 | |||
XM_011545196.2 | XP_011543498.1 | 4 | 586 | |||
XM_011545197.2 | XP_011543499.1 | 4 | 902 | |||
XM_011545198.2 | XP_011543500.1 | 4 | 610 | |||
XM_011545199.2 | XP_011543501.1 | 4 | 588 | |||
XM_011545201.2 | XP_011543503.1 | 4 | 593 | |||
GenBank mRNA | AF084557.1 | - | 3 | 169 | ||
BC022239.1 | - | 4 | 531 | 4390573 | ||
BC051820.1 | - | 4 | 591 | 5188805 | ||
BQ946081.1 | - | 4 | 557 | |||
DR004757.1 | - | 4 | 617 |