Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pcdha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198117.2 | NP_932785.1 | 1 | 787 | ||
GenBank mRNA | AY013760.1 | - | 1 | 674 | ||
BC030897.1 | - | 1 | 787 | |||
BC141390.1 | - | 1 | 768 | 9056003 |