Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Scn11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019265.2 | NP_062138.1 | 13 | 1816 | ||
XM_008766674.1 | XP_008764896.1 | 13 | 1836 | |||
GenBank mRNA | AF059030.2 | - | 13 | 1816 | ||
AJ237852.1 | - | 12 | 1765 |