Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Itih3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017351.1 | NP_059047.1 | 7 | 765 | ||
XM_006252649.1 | XP_006252711.1 | 7 | 995 | |||
XM_006252650.1 | XP_006252712.1 | 7 | 995 | |||
XM_006252653.1 | XP_006252715.1 | 7 | 995 | |||
XM_008771002.1 | XP_008769224.1 | 7 | 995 | |||
GenBank mRNA | X83231.1 | - | 7 | 765 |