Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Syt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031666.1 | NP_113854.1 | 4 | 1090 | ||
GenBank mRNA | AF375463.1 | - | 4 | 1090 | ||
DQ437524.1 | - | 4 | 1090 |