Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ELOVL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022726.3 | NP_073563.1 | 6 | 1130 | ||
GenBank mRNA | AF277094.1 | - | 6 | 958 | ||
AJ420478.1 | - | 1 | 133 | |||
AK055277.1 | - | 6 | 1112 | |||
AK310625.1 | - | 3 | 686 | |||
AK312511.1 | - | 6 | 1085 | |||
AY037298.1 | - | 6 | 1037 | |||
BC038506.1 | - | 6 | 975 | 5263012 |