Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306089.1 | NP_001293018.1 | 19 | 3163 | ||
NM_007345.3 | NP_031371.3 | 19 | 3354 | |||
XM_005266762.3 | XP_005266819.1 | 19 | 3506 | |||
XM_011526165.2 | XP_011524467.1 | 20 | 3508 | |||
XM_011526166.2 | XP_011524468.1 | 18 | 2980 | |||
XM_011526168.2 | XP_011524470.1 | 19 | 3220 | |||
XM_011526169.2 | XP_011524471.1 | 19 | 3219 | |||
GenBank mRNA | AB208834.1 | - | 11 | 1782 | ||
AF085243.1 | - | 19 | 3354 | |||
AF085244.1 | - | 19 | 3354 | |||
BC140859.1 | - | 19 | 3185 | 9021727 | ||
BX648028.1 | - | 19 | 3163 |