Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Aftph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251532.2 | XP_006251594.1 | 2 | 1566 | ||
XM_006251534.2 | XP_006251596.1 | 2 | 1403 | |||
XM_008770443.1 | XP_008768665.1 | 3 | 1720 | |||
XM_223651.8 | XP_223651.5 | 2 | 1565 |