Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002415.1 | NP_002406.1 | 3 | 435 | ||
GenBank mRNA | AF469046.1 | - | 3 | 338 | ||
AK311929.1 | - | 3 | 430 | |||
BC000447.2 | - | 3 | 419 | 2821346 | ||
BC007676.1 | - | 3 | 403 | 3605756 | ||
BC008914.2 | - | 3 | 389 | 3615896 | ||
BC013976.2 | - | 3 | 434 | 4130651 | ||
BC022414.1 | - | 3 | 436 | 4243461 | ||
BC053376.1 | - | 3 | 372 | 6047427 | ||
CR456520.1 | - | 3 | 358 | |||
CR541651.1 | - | 3 | 338 | |||
L10612.1 | - | 3 | 412 | |||
M25639.1 | - | 3 | 388 | |||
M95775.1 | - | 3 | 312 | |||
Z23063.1 | - | 3 | 338 |