Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RPL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000987.3 | NP_000978.1 | 3 | 371 | ||
NM_001315530.1 | NP_001302459.1 | 3 | 664 | |||
NM_001315531.1 | NP_001302460.1 | 3 | 583 | |||
GenBank mRNA | AK311804.1 | - | 3 | 317 | ||
BC071664.1 | - | 3 | 306 | 5189436 | ||
BI091167.1 | - | 3 | 371 | |||
BI596427.1 | - | 3 | 588 | |||
BI603704.1 | - | 3 | 668 | |||
BM928617.1 | - | 2 | 298 | |||
CR456828.1 | - | 2 | 275 |