Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OSBPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015550.3 | NP_056365.1 | 14 | 1892 | ||
NM_145320.2 | NP_663160.1 | 12 | 1472 | |||
NM_145321.2 | NP_663161.1 | 12 | 1457 | |||
NM_145322.2 | NP_663162.1 | 11 | 1364 | |||
NR_104111.1 | 13 | 1565 | ||||
NR_104112.1 | 12 | 1472 | ||||
XM_005249698.3 | XP_005249755.1 | 14 | 1612 | |||
XM_006715681.3 | XP_006715744.1 | 13 | 1786 | |||
XM_006715682.3 | XP_006715745.1 | 13 | 1790 | |||
XM_006715683.3 | XP_006715746.1 | 12 | 1697 | |||
XM_006715684.3 | XP_006715747.1 | 13 | 1549 | |||
XM_011515258.2 | XP_011513560.1 | 14 | 1689 | |||
XM_011515259.2 | XP_011513561.1 | 15 | 2186 | |||
XM_017011948.1 | XP_016867437.1 | 16 | 2049 | |||
XM_017011949.1 | XP_016867438.1 | 14 | 2078 | |||
GenBank mRNA | AB014604.2 | - | 14 | 1829 | ||
AF392444.1 | - | 14 | 1801 | |||
AF491781.1 | - | 13 | 1519 | |||
AF491782.1 | - | 12 | 1426 | |||
AF491783.1 | - | 12 | 1411 | |||
AF491784.1 | - | 11 | 1318 | |||
AF491785.1 | - | 13 | 1519 | |||
AF491786.1 | - | 12 | 1426 | |||
AF515639.1 | - | 6 | 571 | |||
AF515640.1 | - | 5 | 478 | |||
AL832293.1 | - | 11 | 1128 | |||
AY008372.1 | - | 14 | 1722 | |||
BC017731.1 | - | 14 | 1821 | 3909164 |